COM964463853 Combi-Blocks, Inc.

1-Phenyl-2,5,8,11,14,17,20-heptaoxadocosan-22-ol

  • MDL number MFCD24682795
  • Cas Number : 423763-19-3
  • Molecular Weight : 416.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2378

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2378
Availability details for
Quantity
  • Availability
  • Add to Cart