COM964463889 Combi-Blocks, Inc.

3-(2-(2-Methyl-1h-imidazol-1-yl)acetyl)benzonitrile

  • MDL number MFCD30472150
  • Cas Number : 431040-28-7
  • Molecular Weight : 225.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2454

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2454
Availability details for
Quantity
  • Availability
  • Add to Cart