COM964463956 Combi-Blocks, Inc.

(S)-2-Amino-n-(3-methoxyphenyl)-3-phenylpropanamide

  • MDL number MFCD09725246
  • Cas Number : 1217714-95-8
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2639

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2639
Availability details for
Quantity
  • Availability
  • Add to Cart