COM964463959 Combi-Blocks, Inc.

(4-(Benzyloxy)-5-bromo-3-methoxypyridin-2-yl)methanol

  • MDL number MFCD29917011
  • Cas Number : 895134-17-5
  • Molecular Weight : 324.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2645

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2645
Availability details for
Quantity
  • Availability
  • Add to Cart