COM964463984 Combi-Blocks, Inc.

5-Nitro-1h-1,2,4-triazole

  • MDL number MFCD09842321
  • Cas Number : 24807-55-4
  • Molecular Weight : 114.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2733
Availability details for
Quantity
  • Availability
  • Add to Cart