COM964463992 Combi-Blocks, Inc.

(S,E)-2-Amino-3-phenyl-n-styrylpropanamide

  • MDL number MFCD30471425
  • Cas Number : 1449688-89-4
  • Molecular Weight : 266.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2754
Availability details for
Quantity
  • Availability
  • Add to Cart