COM964464009 Combi-Blocks, Inc.

4-(Hydrazonomethyl)phenol

  • MDL number MFCD00710739
  • Cas Number : 52211-82-2
  • Molecular Weight : 136.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2788

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2788
Availability details for
Quantity
  • Availability
  • Add to Cart