COM964464040 Combi-Blocks, Inc.

3-[(Diphenylmethylene)amino]dihydrofuran-2(3h)-one

  • MDL number MFCD29047108
  • Cas Number : 88539-65-5
  • Molecular Weight : 263.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2865

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2865
Availability details for
Quantity
  • Availability
  • Add to Cart