COM964464058 Combi-Blocks, Inc.

Disodium mono(1,3-dihydroxypropan-2-yl phosphate) mono(2,3-dihydroxypropyl phosphate) hydrate

  • MDL number MFCD30478747
  • Molecular Weight : 406.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2911

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2911
Availability details for
Quantity
  • Availability
  • Add to Cart