COM964464069 Combi-Blocks, Inc.

Fluoroimide

  • MDL number MFCD00053087
  • Cas Number : 41205-21-4
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2937

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2937
Availability details for
Quantity
  • Availability
  • Add to Cart