COM964464073 Combi-Blocks, Inc.

1-(Carboxymethyl)-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD20646967
  • Cas Number : 1379181-19-7
  • Molecular Weight : 197.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2943

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-2943
Availability details for
Quantity
  • Availability
  • Add to Cart