COM964464109 Combi-Blocks, Inc.

Fmoc-gln(trt)-thr(psime,mepro)-oh

  • MDL number MFCD18427356
  • Cas Number : 1572725-72-4
  • Molecular Weight : 751.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3028

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3028
Availability details for
Quantity
  • Availability
  • Add to Cart