COM964464116 Combi-Blocks, Inc.

2-(3-Cyano-4-methyl-5-phenyl-5-(trifluoromethyl)furan-2(5h)-ylidene)malononitrile

  • MDL number MFCD29917027
  • Cas Number : 436097-14-2
  • Molecular Weight : 315.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3048

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3048
Availability details for
Quantity
  • Availability
  • Add to Cart