COM964464183 Combi-Blocks, Inc.

2-Methyl-4-(piperidin-4-yl)pyrimidine

  • MDL number MFCD10700265
  • Cas Number : 949100-33-8
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3238

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3238
Availability details for
Quantity
  • Availability
  • Add to Cart