COM964464218 Combi-Blocks, Inc.

(R)-4-Boc-5-methyl-1,4-diazepane 1/2 dbta salt

  • MDL number MFCD30471579
  • Molecular Weight : 786.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3353

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3353
Availability details for
Quantity
  • Availability
  • Add to Cart