COM964464244 Combi-Blocks, Inc.

tert-Butyl 3-(aminomethyl)piperidine-1-carboxylate hydrochloride

  • MDL number MFCD06738968
  • Cas Number : 1029689-80-2
  • Molecular Weight : 250.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3423

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3423
Availability details for
Quantity
  • Availability
  • Add to Cart