COM964464254 Combi-Blocks, Inc.

1-(4-[(Trifluoromethyl)sulfanyl]phenyl)ethan-1-amine

  • MDL number MFCD18703057
  • Cas Number : 871519-25-4
  • Molecular Weight : 221.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3449

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3449
Availability details for
Quantity
  • Availability
  • Add to Cart