COM964464275 Combi-Blocks, Inc.

Fmoc-l-lys(octanoyl)-oh

  • MDL number MFCD24849706
  • Cas Number : 1128181-16-7
  • Molecular Weight : 494.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3511

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3511
Availability details for
Quantity
  • Availability
  • Add to Cart