COM964464306 Combi-Blocks, Inc.

2,2-Dimethyl-1-morpholinopropane-1-thione

  • MDL number MFCD30471954
  • Cas Number : 2007920-61-6
  • Molecular Weight : 187.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3596
Availability details for
Quantity
  • Availability
  • Add to Cart