COM964464307 Combi-Blocks, Inc.

3-(4-Piperidinyl)-1,2-benzisoxazole hydrochloride

  • MDL number MFCD27978162
  • Cas Number : 84163-22-4
  • Molecular Weight : 238.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3598
Availability details for
Quantity
  • Availability
  • Add to Cart