COM964464311 Combi-Blocks, Inc.

2-Methyl-3-(triethylsilyloxy)propyl methanesulfonate

  • MDL number MFCD29067778
  • Cas Number : 718633-39-7
  • Molecular Weight : 282.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3609

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3609
Availability details for
Quantity
  • Availability
  • Add to Cart