COM964464339 Combi-Blocks, Inc.

(3,4-Dimethoxyphenyl)(4-methoxyphenyl)methanone

  • MDL number MFCD00088740
  • Cas Number : 2898-54-6
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3682

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3682
Availability details for
Quantity
  • Availability
  • Add to Cart