COM964464347 Combi-Blocks, Inc.

(2R,4S)-6-Fluoro-2-methylspiro[2,3-dihydrochromene-4,5'-imidazolidine]-2',4'-dione

  • MDL number MFCD19105458
  • Cas Number : 102916-95-0
  • Molecular Weight : 250.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3704

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3704
Availability details for
Quantity
  • Availability
  • Add to Cart