COM964464356 Combi-Blocks, Inc.

1,8-Bis(methoxymethoxy)naphthalene

  • MDL number MFCD27756710
  • Cas Number : 172915-62-7
  • Molecular Weight : 248.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3725

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3725
Availability details for
Quantity
  • Availability
  • Add to Cart