COM964464446 Combi-Blocks, Inc.

2-(2-Fluorobenzyl)-1,5-dimethyl-4-nitro-1h-pyrazol-3(2h)-one

  • MDL number MFCD30472143
  • Cas Number : 2007909-46-6
  • Molecular Weight : 265.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3998

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-3998
Availability details for
Quantity
  • Availability
  • Add to Cart