COM964464449 Combi-Blocks, Inc.

(S)-Methyl 5-(methylthio)-3,4-dihydro-2h-pyrrole-2-carboxylate

  • MDL number MFCD30471873
  • Cas Number : 915719-88-9
  • Molecular Weight : 173.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4014

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4014
Availability details for
Quantity
  • Availability
  • Add to Cart