COM964464460 Combi-Blocks, Inc.

(E)-2-(Benzylideneamino)-1,2-dihydroisoquinoline-3,4-dione

  • MDL number MFCD24848690
  • Cas Number : 1432075-70-1
  • Molecular Weight : 264.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4036

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4036
Availability details for
Quantity
  • Availability
  • Add to Cart