COM964464498 Combi-Blocks, Inc.

3,3-Dimethyl-4-nitrobutanoic acid

  • MDL number MFCD30471719
  • Cas Number : 1936653-09-6
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4130

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4130
Availability details for
Quantity
  • Availability
  • Add to Cart