COM964464514 Combi-Blocks, Inc.

Cis-1-(tert-butoxycarbonyl)-4-(ethoxycarbonyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD30472107
  • Cas Number : 1212409-03-4
  • Molecular Weight : 287.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4175

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4175
Availability details for
Quantity
  • Availability
  • Add to Cart