COM964464524 Combi-Blocks, Inc.

4-Cyano-4'-n-butyl-p-terphenyl

  • MDL number MFCD08276457
  • Cas Number : 66044-94-8
  • Molecular Weight : 311.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4216

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4216
Availability details for
Quantity
  • Availability
  • Add to Cart