COM964464525 Combi-Blocks, Inc.

2-(Trifluoromethyl)benzo[d]thiazol-5-amine

  • MDL number MFCD18821454
  • Cas Number : 58460-24-5
  • Molecular Weight : 218.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4221

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4221
Availability details for
Quantity
  • Availability
  • Add to Cart