COM964464542 Combi-Blocks, Inc.

1,11-Dihydro-8-methyl-2h-pyrimido[4,5-a]carbazol-2-one

  • MDL number MFCD30471481
  • Cas Number : 248246-44-8
  • Molecular Weight : 249.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4273

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4273
Availability details for
Quantity
  • Availability
  • Add to Cart