COM964464563 Combi-Blocks, Inc.

(3R,5R)-3,5-Dimethyl-4-phenylpiperidin-4-ol

  • MDL number MFCD30472099
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4328

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4328
Availability details for
Quantity
  • Availability
  • Add to Cart