COM964464614 Combi-Blocks, Inc.

(1S,4R)-Methyl 4-aminocyclopent-2-enecarboxylate

  • MDL number MFCD16293378
  • Cas Number : 138923-03-2
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4472
Availability details for
Quantity
  • Availability
  • Add to Cart