COM964464626 Combi-Blocks, Inc.

(3R)-6,7-Dichloro-2,3-dihydrobenzo[b]furan-3-ylamine

  • MDL number MFCD14562541
  • Cas Number : 1213494-33-7
  • Molecular Weight : 204.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4499
Availability details for
Quantity
  • Availability
  • Add to Cart