COM964464633 Combi-Blocks, Inc.

(1S,2S)-2-(tert-Butoxycarbonyl)cyclohexanecarboxylic acid

  • MDL number MFCD28975315
  • Cas Number : 488703-60-2
  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4523

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4523
Availability details for
Quantity
  • Availability
  • Add to Cart