COM964464638 Combi-Blocks, Inc.

N-Methyl-1-(1-methyl-1,2,3,4-tetrahydroquinolin-6-yl)methanamine

  • MDL number MFCD08060948
  • Cas Number : 916792-22-8
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4529

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4529
Availability details for
Quantity
  • Availability
  • Add to Cart