COM964464682 Combi-Blocks, Inc.

Benzyl (2-oxocyclohexyl)carbamate

  • MDL number MFCD08061946
  • Cas Number : 31236-61-0
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4625

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4625
Availability details for
Quantity
  • Availability
  • Add to Cart