COM964464700 Combi-Blocks, Inc.

Potassium 5-bromo-6-chloro-1h-indol-3-yl sulfate

  • MDL number MFCD00216685
  • Cas Number : 6581-24-4
  • Molecular Weight : 364.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4676

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4676
Availability details for
Quantity
  • Availability
  • Add to Cart