COM964464738 Combi-Blocks, Inc.

2'-Chloro-4-dimethylaminoazobenzene

  • MDL number MFCD00059478
  • Cas Number : 3010-47-7
  • Molecular Weight : 259.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4754
Availability details for
Quantity
  • Availability
  • Add to Cart