COM964464744 Combi-Blocks, Inc.

3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(((benzyloxy)carbonyl)amino)propanoic acid

  • MDL number MFCD28015810
  • Cas Number : 1822422-91-2
  • Molecular Weight : 460.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4767

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4767
Availability details for
Quantity
  • Availability
  • Add to Cart