COM964464746 Combi-Blocks, Inc.

1-(4-Chlorophenoxy)-3,3-dimethylbutan-2-one

  • MDL number MFCD00459124
  • Cas Number : 24473-06-1
  • Molecular Weight : 226.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4773

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4773
Availability details for
Quantity
  • Availability
  • Add to Cart