COM964464810 Combi-Blocks, Inc.

3-Cyclohexene-1-carboxylic acid, 6-[[(1,1-dimethylethoxy)carbonyl]amino]-, methyl ester, (1r,6s)-rel-

  • MDL number MFCD28964068
  • Cas Number : 836607-50-2
  • Molecular Weight : 255.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4959
Availability details for
Quantity
  • Availability
  • Add to Cart