COM964464823 Combi-Blocks, Inc.

2-Nitro-6-(trifluoromethoxy)anisole

  • MDL number MFCD18399917
  • Cas Number : 1261764-67-3
  • Molecular Weight : 237.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4980
Availability details for
Quantity
  • Availability
  • Add to Cart