COM964464824 Combi-Blocks, Inc.

1-(Isopropoxycarbonyl)-2,3-dihydro-1h-pyrrolizine-7-carboxylic acid

  • MDL number MFCD30294609
  • Cas Number : 66635-70-9
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4986

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-4986
Availability details for
Quantity
  • Availability
  • Add to Cart