COM964464846 Combi-Blocks, Inc.

(2,3-Dichloro-6-nitrophenyl)-hydrazine

  • MDL number MFCD30471402
  • Cas Number : 861363-57-7
  • Molecular Weight : 222.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5029

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5029
Availability details for
Quantity
  • Availability
  • Add to Cart