COM964464892 Combi-Blocks, Inc.

1-Bromo-3-(prop-1-yn-1-yl)benzene

  • MDL number MFCD17677178
  • Cas Number : 66952-36-1
  • Molecular Weight : 195.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5159
Availability details for
Quantity
  • Availability
  • Add to Cart