COM964464935 Combi-Blocks, Inc.

Ethyl 2,2-difluorohex-3-enoate

  • MDL number MFCD23704335
  • Cas Number : 1346523-54-3
  • Molecular Weight : 178.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5304

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5304
Availability details for
Quantity
  • Availability
  • Add to Cart