COM964464959 Combi-Blocks, Inc.

Methyl 2-amino-5-bromo-3-methoxybenzoate

  • MDL number MFCD20040042
  • Cas Number : 115378-21-7
  • Molecular Weight : 260.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5361

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5361
Availability details for
Quantity
  • Availability
  • Add to Cart