COM964464969 Combi-Blocks, Inc.

(1-(2,6-Dichlorophenyl)-4-isopropyl-1h-1,2,3-triazol-5-yl)methanol

  • MDL number MFCD11046173
  • Cas Number : 959800-67-0
  • Molecular Weight : 286.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5391

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QV-5391
Availability details for
Quantity
  • Availability
  • Add to Cart